Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNASEL
Entrez Gene ID
715647
Gene Name
ribonuclease L
Gene Aliases
-
Chromosome Location
Chr.1: 39825920 - 39841870 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042433.2 NP_001035898.1 - 1470 64
XM_015121466.2 XP_014976952.1 - 1744 64
GenBank mRNA DQ644021.1 - - 1473 64
EF467993.1 - - 1472 64
JU470652.1 - - 1641 64

View Full Product Details