Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LPA
Entrez Gene ID
709528
Gene Name
lipoprotein(a)
Gene Aliases
Apo(a), Lp(a)
Chromosome Location
Chr.4: 9990597 - 10143743 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028847001.1 XP_028702834.1 - 3898 69
GenBank mRNA J04635.1 - - 1984 69

View Full Product Details