Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MBP-A
Entrez Gene ID
707965
Gene Name
mannose-binding protein A
Gene Aliases
MBL1
Chromosome Location
Chr.9: 58198614 - 58203290 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001096399.4 XP_001096399.1 - 489 62
GenBank mRNA L43912.1 - - 364 62

View Full Product Details