Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSTM3
Entrez Gene ID
700303
Gene Name
glutathione S-transferase mu 3
Gene Aliases
EGK_01059
Chromosome Location
Chr.1: 114389612 - 114392970 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261580.1 NP_001248509.1 - 101 110
GenBank mRNA JU322637.1 - - 101 110
KC734500.1 - - 46 110

View Full Product Details