Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARL6IP6
Entrez Gene ID
695619
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 6
Gene Aliases
EGK_04443
Chromosome Location
Chr.12: 39838925 - 39883510 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260607.1 NP_001247536.1 - 875 129
XM_028830405.1 XP_028686238.1 - 645 129
GenBank mRNA JU474439.1 - - 624 129
JV046271.1 - - 875 129
JV635735.1 - - 1278 129

View Full Product Details