Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDO1
Entrez Gene ID
696387
Gene Name
cysteine dioxygenase type 1
Gene Aliases
-
Chromosome Location
Chr.6: 113344420 - 113356166 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260608.1 NP_001247537.1 - 585 67
XM_028849098.1 XP_028704931.1 - 513 67
GenBank mRNA JU321610.1 - - 651 67
JV045144.1 - - 585 67
JV601372.1 - - 593 67

View Full Product Details