Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDHAF2
Entrez Gene ID
693401
Gene Name
succinate dehydrogenase complex assembly factor 2
Gene Aliases
C14H11orf79, EGK_06141
Chromosome Location
Chr.14: 12729251 - 12748142 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193692.1 NP_001180621.1 - 353 87
XM_015114004.2 XP_014969490.1 - 517 87
XM_015114005.2 XP_014969491.1 - 315 87
XM_028832505.1 XP_028688338.1 - 374 87
GenBank mRNA JU321399.1 - - 283 87
JU473320.1 - - 285 87
JV047471.1 - - 288 87
JV472769.1 - - 282 87

View Full Product Details