Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COIL
Entrez Gene ID
708103
Gene Name
coilin
Gene Aliases
-
Chromosome Location
Chr.16: 39852573 - 39873201 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257478.1 NP_001244407.1 - 1440 88
GenBank mRNA JU336548.1 - - 1440 88

View Full Product Details