Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VSIG4
Entrez Gene ID
709685
Gene Name
V-set and immunoglobulin domain containing 4
Gene Aliases
-
Chromosome Location
Chr.X: 62624541 - 62643140 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001099264.2 XP_001099264.2 - 176 84
XM_015127593.2 XP_014983079.1 - 176 84
XM_015127594.1 XP_014983080.1 - 176 84
XM_015127595.2 XP_014983081.1 - 176 84
XM_015127596.1 XP_014983082.1 - 181 84
XM_028841902.1 XP_028697735.1 - 176 84
XM_028841903.1 XP_028697736.1 - 176 84

View Full Product Details