Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SSR3
Entrez Gene ID
706518
Gene Name
signal sequence receptor subunit 3
Gene Aliases
EGK_11971
Chromosome Location
Chr.2: 58842944 - 58856705 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266544.1 NP_001253473.1 - 570 83
GenBank mRNA JU320285.1 - - 557 83
JU471158.1 - - 570 83
JV044116.1 - - 570 83
JV518108.1 - - 519 83

View Full Product Details