Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APMAP
Entrez Gene ID
704948
Gene Name
adipocyte plasma membrane associated protein
Gene Aliases
C10H20orf3, C20orf3, EGK_02740
Chromosome Location
Chr.10: 41635490 - 41663367 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266050.1 NP_001252979.1 - 160 60
XM_028827285.1 XP_028683118.1 - 331 60
GenBank mRNA JU321400.1 - - 160 60
JV513791.1 - - 62 60
JV656575.1 - - 178 60

View Full Product Details