Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNAPIN
Entrez Gene ID
715546
Gene Name
SNAP associated protein
Gene Aliases
EGK_01334
Chromosome Location
Chr.1: 97403919 - 97406968 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257650.1 NP_001244579.1 - 376 80
GenBank mRNA JU333731.1 - - 376 80
JV046610.1 - - 334 80
JV462926.1 - - 261 80
JV647038.1 - - 376 80

View Full Product Details