Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NT5C3A
Entrez Gene ID
708743
Gene Name
5'-nucleotidase, cytosolic IIIA
Gene Aliases
EGK_13788, NT5C3
Chromosome Location
Chr.3: 82015693 - 82065603 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261493.1 NP_001248422.1 - 441 122
XM_015133735.2 XP_014989221.1 - 960 122
XM_015133736.2 XP_014989222.1 - 816 122
GenBank mRNA JU334836.1 - - 393 122
JU472937.1 - - 441 122

View Full Product Details