Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPINT2
Entrez Gene ID
714755
Gene Name
serine peptidase inhibitor, Kunitz type 2
Gene Aliases
EGK_10558
Chromosome Location
Chr.19: 37499021 - 37532820 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194343.2 NP_001181272.1 - 959 81
GenBank mRNA JU323207.1 - - 1005 81
JU471549.1 - - 940 81
JV045400.1 - - 929 81
JV600534.1 - - 1033 81
JV637676.1 - - 977 81

View Full Product Details