Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GBP3
- Entrez Gene ID
-
694283
- Gene Name
- guanylate binding protein 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 135415353 - 135430762 on Build Mmul_10
- Gene Symbol
- LOC694538
- Entrez Gene ID
-
694538
- Gene Name
- guanylate-binding protein 1
- Gene Aliases
-
GBP1
- Chromosome Location
-
Chr.1: 135369960 - 135385501 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JU475139.1 |
- |
- |
401 |
157 |
View Full Product Details