Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTGES3
Entrez Gene ID
713353
Gene Name
prostaglandin E synthase 3
Gene Aliases
-
Chromosome Location
Chr.11: 56240089 - 56266116 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JU319709.1 - - 47 124
JU323280.1 - - 47 124
JU472853.1 - - 53 124
JU474053.1 - - 53 124

View Full Product Details