Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALX4
Entrez Gene ID
715652
Gene Name
ALX homeobox 4
Gene Aliases
-
Chromosome Location
Chr.14: 21435830 - 21485909 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001113643.4 XP_001113643.1 - 1111 118

View Full Product Details