Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDH20
Entrez Gene ID
696607
Gene Name
protocadherin 20
Gene Aliases
-
Chromosome Location
Chr.17: 40143646 - 40150528 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028837500.1 XP_028693333.1 - 816 110

View Full Product Details