Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIS12
Entrez Gene ID
712202
Gene Name
MIS12 kinetochore complex component
Gene Aliases
-
Chromosome Location
Chr.16: 5282493 - 5287159 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194189.2 NP_001181118.1 - 2118 152
XM_001101672.4 XP_001101672.1 - 2161 152
XM_015118306.2 XP_014973792.1 - 2117 152

View Full Product Details