Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB3
Entrez Gene ID
722240
Gene Name
zinc finger and BTB domain containing 3
Gene Aliases
-
Chromosome Location
Chr.14: 11442217 - 11448890 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015113960.2 XP_014969446.2 - 230 81
GenBank mRNA JV661755.1 - - 193 81

View Full Product Details