Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
AGTR1
Entrez Gene ID
712773
Gene Name
angiotensin II receptor type 1
Gene Aliases
EGK_12019
Chromosome Location
Chr.2: 50825360 - 50870238 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001110548.4 XP_001110548.1 - 221 143
XM_015131970.2 XP_014987456.1 - 220 143

View Full Product Details