Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTOS
Entrez Gene ID
698202
Gene Name
otospiralin
Gene Aliases
EGK_04973
Chromosome Location
Chr.12: 127518588 - 127520577 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260881.1 NP_001247810.1 - 250 147
XM_015111494.2 XP_014966980.1 - 511 147
GenBank mRNA JU322652.1 - - 250 147
JV567751.1 - - 248 147

View Full Product Details