Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNJ1
Entrez Gene ID
714848
Gene Name
potassium inwardly rectifying channel subfamily J member 1
Gene Aliases
EGK_07084
Chromosome Location
Chr.14: 121558025 - 121629306 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001113433.3 XP_001113433.1 - 190 153

View Full Product Details