Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNG4
Entrez Gene ID
715185
Gene Name
potassium voltage-gated channel modifier subfamily G member 4
Gene Aliases
-
Chromosome Location
Chr.20: 71233033 - 71251737 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015126715.2 XP_014982201.2 - 1262 132

View Full Product Details