Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLA2G5
Entrez Gene ID
704629
Gene Name
phospholipase A2 group V
Gene Aliases
-
Chromosome Location
Chr.1: 204642267 - 204675305 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261213.1 NP_001248142.1 - 256 125
GenBank mRNA JU335014.1 - - 256 125

View Full Product Details