Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR4D11
Entrez Gene ID
699579
Gene Name
olfactory receptor, family 4, subfamily D, member 11
Gene Aliases
EGK_06186
Chromosome Location
Chr.14: 14468125 - 14469060 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193875.1 NP_001180804.1 - 248 102

View Full Product Details