Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLX4IP
Entrez Gene ID
718563
Gene Name
SLX4 interacting protein
Gene Aliases
-
Chromosome Location
Chr.10: 55916261 - 56115971 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001116207.3 XP_001116207.2 - 407 138
GenBank mRNA JU332918.1 - - 283 138
JU474321.1 - - 280 138

View Full Product Details