Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ICE2
Entrez Gene ID
706347
Gene Name
interactor of little elongation complex ELL subunit 2
Gene Aliases
NARG2
Chromosome Location
Chr.7: 36884696 - 36940821 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015142361.2 XP_014997847.1 - 633 92
XM_015142362.2 XP_014997848.1 - 677 92
XM_015142365.2 XP_014997851.1 - 489 92
XM_015142366.2 XP_014997852.1 - 517 92
XM_015142368.2 XP_014997854.1 - 633 92
GenBank mRNA JU322771.1 - - 439 92
JU477218.1 - - 617 92
JV723423.1 - - 429 92

View Full Product Details