Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHI3L2
Entrez Gene ID
705024
Gene Name
chitinase 3 like 2
Gene Aliases
-
Chromosome Location
Chr.1: 112917797 - 112932579 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015148162.2 XP_015003648.2 - 1620 90
GenBank mRNA JU332724.1 - - 1199 90
JU479402.1 - - 1102 90

View Full Product Details