Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35F1
Entrez Gene ID
714965
Gene Name
solute carrier family 35 member F1
Gene Aliases
-
Chromosome Location
Chr.4: 21428088 - 21827448 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266534.1 NP_001253463.1 - 1115 75
XM_015137311.2 XP_014992797.1 - 1504 75
GenBank mRNA JU333419.1 - - 1115 75
JV574699.1 - - 1180 75

View Full Product Details