Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDNF
Entrez Gene ID
697138
Gene Name
cerebral dopamine neurotrophic factor
Gene Aliases
EGK_19470
Chromosome Location
Chr.9: 15964272 - 15982796 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193826.2 NP_001180755.1 - 201 118

View Full Product Details