Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VSTM4
Entrez Gene ID
710593
Gene Name
V-set and transmembrane domain containing 4
Gene Aliases
C10orf72
Chromosome Location
Chr.9: 48250218 - 48356516 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194390.2 NP_001181319.1 - 481 131
XM_028853465.1 XP_028709298.1 - 499 131
GenBank mRNA JU326897.1 - - 397 131
JV053978.1 - - 439 131
JV644558.1 - - 451 131

View Full Product Details