Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCN1B
Entrez Gene ID
707134
Gene Name
sodium voltage-gated channel beta subunit 1
Gene Aliases
-
Chromosome Location
Chr.19: 34450401 - 34460675 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258167.1 NP_001245096.1 - 475 112
XM_015123520.2 XP_014979006.1 - 1081 112
XM_028838435.1 XP_028694268.1 - 811 112
XM_028838436.1 XP_028694269.1 - 1131 112
XM_028838437.1 XP_028694270.1 - 810 112
GenBank mRNA JU321213.1 - - 587 112
JU473653.1 - - 475 112
JV667885.1 - - 589 112

View Full Product Details