Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAQR3
Entrez Gene ID
697120
Gene Name
progestin and adipoQ receptor family member 3
Gene Aliases
-
Chromosome Location
Chr.5: 54813791 - 54841145 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257693.1 NP_001244622.1 - 658 90
GenBank mRNA JU320618.1 - - 658 90
JU474452.1 - - 540 90
JV044586.1 - - 663 90

View Full Product Details