Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPN20
Entrez Gene ID
711276
Gene Name
protein tyrosine phosphatase non-receptor type 20
Gene Aliases
EGK_19635, PTPN20A
Chromosome Location
Chr.9: 49239796 - 49340960 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001109336.4 XP_001109336.1 - 394 178
XM_015146962.2 XP_015002448.1 - 882 178
XM_015146965.2 XP_015002451.1 - 2140 178
XM_015146968.2 XP_015002454.1 - 304 178
XM_028826354.1 XP_028682187.1 - 844 178
XM_028826355.1 XP_028682188.1 - 945 178
XM_028826356.1 XP_028682189.1 - 447 178
XM_028826357.1 XP_028682190.1 - 752 178
XM_028826358.1 XP_028682191.1 - 832 178
XM_028826359.1 XP_028682192.1 - 304 178

View Full Product Details