Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP83
Entrez Gene ID
711408
Gene Name
centrosomal protein 83
Gene Aliases
CCDC41, EGK_04062
Chromosome Location
Chr.11: 94115879 - 94263098 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015152449.2 XP_015007935.1 - 324 102

View Full Product Details