Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFRSF17
Entrez Gene ID
712212
Gene Name
TNF receptor superfamily member 17
Gene Aliases
EGK_12514
Chromosome Location
Chr.20: 11938898 - 11941907 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001106892.4 XP_001106892.1 - 487 142

View Full Product Details