Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC695533
- Entrez Gene ID
-
695533
- Gene Name
- leukocyte-associated immunoglobulin-like receptor 1
- Gene Aliases
-
LAIR1
- Chromosome Location
-
Chr.19: 54162560 - 54173402 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
KT344776.1 |
- |
- |
479 |
109 |
View Full Product Details