Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP3K11
Entrez Gene ID
716038
Gene Name
mitogen-activated protein kinase kinase kinase 11
Gene Aliases
EGK_06004
Chromosome Location
Chr.14: 8910873 - 8927847 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257468.1 NP_001244397.1 - 2096 147
GenBank mRNA JU321005.1 - - 2096 147
JU473336.1 - - 2094 147
JV044425.1 - - 2008 147

View Full Product Details