Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPP1R8
Entrez Gene ID
716496
Gene Name
protein phosphatase 1 regulatory subunit 8
Gene Aliases
EGK_00448
Chromosome Location
Chr.1: 196871240 - 196892619 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266462.1 NP_001253391.1 - 704 97
XM_028845627.1 XP_028701460.1 - 509 97
GenBank mRNA JU319859.1 - - 733 97
JV045215.1 - - 704 97
JV654991.1 - - 747 97

View Full Product Details