Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ST3GAL1
Entrez Gene ID
698004
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 1
Gene Aliases
EGK_19292
Chromosome Location
Chr.8: 133740173 - 133885675 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261634.1 NP_001248563.1 - 1626 76
XM_015146017.2 XP_015001503.1 - 2191 76
XM_015146018.2 XP_015001504.1 - 2334 76
XM_015146019.2 XP_015001505.1 - 2757 76
XM_028852132.1 XP_028707965.1 - 2614 76
XM_028852133.1 XP_028707966.1 - 2610 76
XM_028852134.1 XP_028707967.1 - 2482 76
XM_028852135.1 XP_028707968.1 - 2535 76
XM_028852136.1 XP_028707969.1 - 2494 76
GenBank mRNA JU473118.1 - - 1829 76
JU473119.1 - - 1626 76

View Full Product Details