Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUV39H1
Entrez Gene ID
711040
Gene Name
suppressor of variegation 3-9 homolog 1
Gene Aliases
EGK_20467
Chromosome Location
Chr.X: 48694681 - 48708159 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261327.1 NP_001248256.1 - 70 89
GenBank mRNA JU473137.1 - - 70 89

View Full Product Details