Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF1AD
Entrez Gene ID
715063
Gene Name
eukaryotic translation initiation factor 1A domain containing
Gene Aliases
EGK_05986
Chromosome Location
Chr.14: 8548566 - 8552068 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194525.1 NP_001181454.1 - 422 110
XM_001111740.4 XP_001111740.1 - 580 110
XM_015113651.2 XP_014969137.1 - 735 110
XM_015113653.2 XP_014969139.1 - 711 110
GenBank mRNA JU322114.1 - - 160 110
JU478357.1 - - 366 110
JV522836.1 - - 324 110

View Full Product Details