Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYYR1
Entrez Gene ID
708008
Gene Name
cysteine and tyrosine rich 1
Gene Aliases
-
Chromosome Location
Chr.3: 20096733 - 20202867 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001104127.4 XP_001104127.2 - 556 82
XM_015133067.2 XP_014988553.2 - 556 82
GenBank mRNA JU331147.1 - - 46 82
JV493995.1 - - 84 82

View Full Product Details