Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PANX3
Entrez Gene ID
711728
Gene Name
pannexin 3
Gene Aliases
EGK_07042
Chromosome Location
Chr.14: 117390645 - 117397237 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001109098.4 XP_001109098.1 - 698 99

View Full Product Details