Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GIPC3
Entrez Gene ID
713364
Gene Name
GIPC PDZ domain containing family member 3
Gene Aliases
-
Chromosome Location
Chr.19: 3474796 - 3478481 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194162.2 NP_001181091.1 - 829 103

View Full Product Details