Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM189A2
Entrez Gene ID
701247
Gene Name
family with sequence similarity 189 member A2
Gene Aliases
C15H9orf61
Chromosome Location
Chr.15: 84774221 - 84839033 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015117864.2 XP_014973350.2 - 1157 179
GenBank mRNA JU337166.1 - - 671 179
JU473647.1 - - 643 179

View Full Product Details