Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DGCR6L
- Entrez Gene ID
-
693512
- Gene Name
- DiGeorge syndrome critical region gene 6 like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 32308023 - 32313689 on Build Mmul_10
- Gene Symbol
- DGCR6
- Entrez Gene ID
-
114669725
- Gene Name
- DiGeorge syndrome critical region gene 6
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 33730003 - 33744079 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV667290.1 |
- |
- |
423 |
116 |
View Full Product Details