Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CELA3B
Entrez Gene ID
574236
Gene Name
chymotrypsin like elastase 3B
Gene Aliases
ELA1, ELA3B
Chromosome Location
Chr.1: 202772569 - 202784313 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195397.3 NP_001182326.2 - 913 132

View Full Product Details