Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL41
Entrez Gene ID
706047
Gene Name
kelch like family member 41
Gene Aliases
EGK_04509, KBTBD10
Chromosome Location
Chr.12: 56608090 - 56626160 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001104263.4 XP_001104263.1 - 1604 176
XM_015110295.2 XP_014965781.1 - 1604 176

View Full Product Details